Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142605.1 | 3905 | UTR 3 | NP_001136077.1 | ||
NM_001258353.1 | 3905 | UTR 3 | NP_001245282.1 | ||
NM_001258354.1 | 3905 | UTR 3 | NP_001245283.1 | ||
NM_004247.3 | 3905 | UTR 3 | NP_004238.3 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271880.1 | 3905 | Intron | NP_001258809.1 | ||
NM_016438.3 | 3905 | Intron | NP_057522.1 | ||
XM_006721946.2 | 3905 | Intron | XP_006722009.1 | ||
XM_006721947.3 | 3905 | Intron | XP_006722010.1 | ||
XM_006721948.3 | 3905 | Intron | XP_006722011.1 | ||
XM_011524891.2 | 3905 | Intron | XP_011523193.1 | ||
XM_017024742.1 | 3905 | Intron | XP_016880231.1 | ||
XM_017024743.1 | 3905 | Intron | XP_016880232.1 |