Product Details
- SNP ID
-
rs7215075
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:53822904 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTGGCCCCAGTCCGGGCCCCGGC[A/G]CGCTAGGCTCACAAAGGCAGGCACA
- Phenotype
-
MIM: 615142
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113521969] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF2B
- Gene Name
- kinesin family member 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032559.4 |
27 |
UTR 5 |
|
|
NP_115948.4 |
View Full Product Details