Product Details

SNP ID
rs34215541
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:111904213 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTGCCTCACCTCGTATCAGTCTGCC[A/C]TGTTATCTTGAATCCATCCAATAAT
Phenotype
MIM: 602874
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UGCG PubMed Links

Gene Details

Gene
UGCG
Gene Name
UDP-glucose ceramide glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003358.2 Intron NP_003349.1
XM_005252186.4 Intron XP_005252243.1
XM_017015107.1 Intron XP_016870596.1

View Full Product Details