Product Details

SNP ID
rs11623680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22922580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCTGTCAGGAAATAATTATGT[C/G]GGTGACAAGGGGCCAGAAGCTCTAG
Phenotype
MIM: 604045
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101926933 PubMed Links

Gene Details

Gene
LOC101926933
Gene Name
uncharacterized LOC101926933
There are no transcripts associated with this gene.

Gene
PRMT5
Gene Name
protein arginine methyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039619.2 Intron NP_001034708.1
NM_001282953.1 Intron NP_001269882.1
NM_001282954.1 Intron NP_001269883.1
NM_001282955.1 Intron NP_001269884.1
NM_001282956.1 Intron NP_001269885.1
NM_006109.4 Intron NP_006100.2
XM_017020927.1 Intron XP_016876416.1
Gene
PRMT5-AS1
Gene Name
PRMT5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBM23
Gene Name
RNA binding motif protein 23
There are no transcripts associated with this gene.

View Full Product Details