Product Details

SNP ID
rs17575205
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:109494376 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGAGCCCCGGGAGACTTGGAGGAGG[C/G]ACCCCAGCAGTAAAGGGGAGACAGA
Phenotype
MIM: 615689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AMIGO1 PubMed Links

Gene Details

Gene
AMIGO1
Gene Name
adhesion molecule with Ig like domain 1
There are no transcripts associated with this gene.

Gene
ATXN7L2
Gene Name
ataxin 7 like 2
There are no transcripts associated with this gene.

Gene
CYB561D1
Gene Name
cytochrome b561 family member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134400.1 Intron NP_001127872.1
NM_001134402.1 Intron NP_001127874.1
NM_001134403.1 Intron NP_001127875.1
NM_001134404.1 Intron NP_001127876.1
NM_182580.2 Intron NP_872386.1
XM_005270775.2 Intron XP_005270832.1
XM_005270776.3 Intron XP_005270833.1
XM_005270777.2 Intron XP_005270834.1
XM_011541286.2 Intron XP_011539588.1
XM_011541287.2 Intron XP_011539589.2
XM_017001078.1 Intron XP_016856567.1
XM_017001079.1 Intron XP_016856568.1

View Full Product Details