Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134400.1 | Intron | NP_001127872.1 | |||
NM_001134402.1 | Intron | NP_001127874.1 | |||
NM_001134403.1 | Intron | NP_001127875.1 | |||
NM_001134404.1 | Intron | NP_001127876.1 | |||
NM_182580.2 | Intron | NP_872386.1 | |||
XM_005270775.2 | Intron | XP_005270832.1 | |||
XM_005270776.3 | Intron | XP_005270833.1 | |||
XM_005270777.2 | Intron | XP_005270834.1 | |||
XM_011541286.2 | Intron | XP_011539588.1 | |||
XM_011541287.2 | Intron | XP_011539589.2 | |||
XM_017001078.1 | Intron | XP_016856567.1 | |||
XM_017001079.1 | Intron | XP_016856568.1 |