Product Details

SNP ID
rs1055702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185502339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCGTGTCATAGCCCTCTGGGGG[C/T]TTTGTGCGGGCTCTTGCGTGGGTTT
Phenotype
MIM: 605889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDLIM3 PubMed Links

Gene Details

Gene
PDLIM3
Gene Name
PDZ and LIM domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114107.4 1030 Silent Mutation AAA,AAG K302K NP_001107579.1
NM_001257962.1 1030 Silent Mutation AAA,AAG K262K NP_001244891.1
NM_001257963.1 1030 Silent Mutation AAA,AAG K183K NP_001244892.1
NM_014476.5 1030 Silent Mutation AAA,AAG K350K NP_055291.2

View Full Product Details