Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301861.1 | Intron | NP_001288790.1 | |||
NM_001301867.1 | Intron | NP_001288796.1 | |||
NM_033254.3 | Intron | NP_150279.1 | |||
XM_005247891.2 | Intron | XP_005247948.1 | |||
XM_005247892.2 | Intron | XP_005247949.1 | |||
XM_005247897.2 | Intron | XP_005247954.1 | |||
XM_011513305.2 | Intron | XP_011511607.1 | |||
XM_017007441.1 | Intron | XP_016862930.1 | |||
XM_017007442.1 | Intron | XP_016862931.1 | |||
XM_017007443.1 | Intron | XP_016862932.1 | |||
XM_017007444.1 | Intron | XP_016862933.1 | |||
XM_017007445.1 | Intron | XP_016862934.1 | |||
XM_017007446.1 | Intron | XP_016862935.1 | |||
XM_017007447.1 | Intron | XP_016862936.1 | |||
XM_017007448.1 | Intron | XP_016862937.1 | |||
XM_017007449.1 | Intron | XP_016862938.1 | |||
XM_017007450.1 | Intron | XP_016862939.1 | |||
XM_017007451.1 | Intron | XP_016862940.1 | |||
XM_017007452.1 | Intron | XP_016862941.1 | |||
XM_017007453.1 | Intron | XP_016862942.1 | |||
XM_017007454.1 | Intron | XP_016862943.1 | |||
XM_017007455.1 | Intron | XP_016862944.1 |