Product Details

SNP ID
rs1381057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121436127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTCATCTATGAACAAACCTGTT[C/T]GGTCACTTTGGAGCATACCCTCTCG
Phenotype
MIM: 604419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLQ PubMed Links

Gene Details

Gene
POLQ
Gene Name
DNA polymerase theta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199420.3 7667 Missense Mutation CAA,CGA Q2513R NP_955452.3
XM_011512343.2 7667 Missense Mutation CAA,CGA Q2467R XP_011510645.1
XM_011512347.2 7667 Missense Mutation CAA,CGA Q2513R XP_011510649.1
XM_011512348.2 7667 Missense Mutation CAA,CGA Q2342R XP_011510650.1
XM_011512352.2 7667 Intron XP_011510654.1
XM_011512354.2 7667 Missense Mutation CAA,CGA Q1750R XP_011510656.1
XM_017005565.1 7667 Missense Mutation CAA,CGA Q2123R XP_016861054.1

View Full Product Details