Product Details
- SNP ID
-
rs11155
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:183097612 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGCCCAGACTGCTCTAAGGGCAAAC[A/G]CGACCCCTTTCCTCTTCTAGGACCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WWC2
PubMed Links
Gene Details
- Gene
- WWC2
- Gene Name
- WW and C2 domain containing 2
There are no transcripts associated with this gene.
- Gene
- WWC2-AS2
- Gene Name
- WWC2 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details