Product Details
- SNP ID
-
rs288038
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138871844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCGAAATGTATTATATAATAGTT[A/G]GAAAGTGTTTTTTTTCATGGTTCCT
- Phenotype
-
MIM: 116805
MIM: 610868
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTNNA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148797416] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTNNA1
- Gene Name
- catenin alpha 1
- Gene
- LRRTM2
- Gene Name
- leucine rich repeat transmembrane neuronal 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015564.2 |
3174 |
UTR 3 |
|
|
NP_056379.1 |
View Full Product Details