Product Details
- SNP ID
-
rs1622154
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:618428 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCACAATAGGACACATGTGTTAA[A/C]GTTTGTCTTTCAATCTTTTTACATA
- Phenotype
-
MIM: 616475
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP72
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78625104] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP72
- Gene Name
- centrosomal protein 72
- Gene
- LOC100996325
- Gene Name
- uncharacterized LOC100996325
There are no transcripts associated with this gene.
View Full Product Details