Product Details

SNP ID
rs920980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:896868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATACTCCATGCCATGTCAGTTCAC[A/G]GGGGGGGTTCTGTTTGTCCATTTGT
Phenotype
MIM: 604507
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD9 PubMed Links

Gene Details

Gene
BRD9
Gene Name
bromodomain containing 9
There are no transcripts associated with this gene.

Gene
TRIP13
Gene Name
thyroid hormone receptor interactor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166260.1 Intron NP_001159732.1
NM_004237.3 Intron NP_004228.1
XM_011514163.2 Intron XP_011512465.1

View Full Product Details