Product Details

SNP ID
rs1045012
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:99386731 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACCATGAGGTGCATATCTATGAAAA[C/G]AGCGGTGCCAAATGGACCAAGGTGC
Phenotype
MIM: 604223 MIM: 607075
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005720.3 231 Missense Mutation AAC,AAG N37K NP_005711.1
XM_006715825.1 231 Missense Mutation AAC,AAG N37K XP_006715888.1
XM_006715826.1 231 Missense Mutation AAC,AAG N37K XP_006715889.1
Gene
PDAP1
Gene Name
PDGFA associated protein 1
There are no transcripts associated with this gene.

View Full Product Details