Product Details

SNP ID
rs849388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:106866400 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTGCAGTATGTCCTATTTATAC[A/G]TAAGTGCTTTGTGAAGGGTAGGTAC
Phenotype
MIM: 601232
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3CG PubMed Links

Gene Details

Gene
PIK3CG
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282426.1 Intron NP_001269355.1
NM_001282427.1 Intron NP_001269356.1
NM_002649.3 Intron NP_002640.2
XM_005250443.3 Intron XP_005250500.1
XM_011516316.1 Intron XP_011514618.1
XM_011516317.1 Intron XP_011514619.1
XM_017012328.1 Intron XP_016867817.1

View Full Product Details