Product Details

SNP ID
rs7600326
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:68291148 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGTAGAACAGTATATGCCAGGAG[C/T]GATCACTTAACGTGCAGATTAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNRIP1 PubMed Links

Gene Details

Gene
CNRIP1
Gene Name
cannabinoid receptor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111101.1 Intron NP_001104571.1
NM_015463.2 Intron NP_056278.1
XM_005264249.1 Intron XP_005264306.1
XM_006711984.1 Intron XP_006712047.1

View Full Product Details