Product Details

SNP ID
rs1129763
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:8056485 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTTTTGCAAAGCTCCACCACAACA[C/T]GCAGACCTTCCAAGGTGGGCAAGAC
Phenotype
MIM: 602565 MIM: 608529
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL25 PubMed Links

Gene Details

Gene
CCL25
Gene Name
C-C motif chemokine ligand 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201359.1 422 Missense Mutation ACG,ATG T104M NP_001188288.1
NM_005624.3 422 Missense Mutation ACG,ATG T104M NP_005615.2
XM_011528177.2 422 Missense Mutation ACG,ATG T104M XP_011526479.1
XM_017027119.1 422 Missense Mutation ACG,ATG T164M XP_016882608.1
XM_017027120.1 422 Missense Mutation ACG,ATG T104M XP_016882609.1
XM_017027121.1 422 Missense Mutation ACG,ATG T104M XP_016882610.1
XM_017027122.1 422 Missense Mutation ACG,ATG T104M XP_016882611.1
Gene
FBN3
Gene Name
fibrillin 3
There are no transcripts associated with this gene.

View Full Product Details