Product Details

SNP ID
rs584307
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:7790444 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGAAGTGGGGGCCCCGGGTCCATCC[C/T]TAGCTTAGATTCCCAGCATCACCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC4GP1 PubMed Links

Gene Details

Gene
CLEC4GP1
Gene Name
C-type lectin domain family 4 member G pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details