Product Details

SNP ID
rs1537318
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:197084264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACGGAGAGCAAAAATCACTTTAC[A/G]TACTCATGATTGGCTTTAATATTTC
Phenotype
MIM: 605481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10751 UTR 3 NP_001193775.1
NM_018136.4 10751 UTR 3 NP_060606.3

View Full Product Details