Product Details
- SNP ID
-
hCV8363175
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41619592 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGGCCGGCCGGGGTAGCTGAGT[C/G]CTCAGCGGGTGGTCTGGTGGACCTG
- Phenotype
-
MIM: 148067
MIM: 148069
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT16
PubMed Links
Gene Details
- Gene
- KRT16
- Gene Name
- keratin 16
There are no transcripts associated with this gene.
- Gene
- KRT17
- Gene Name
- keratin 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000422.2 |
|
Intron |
|
|
NP_000413.1 |
- Gene
- KRT42P
- Gene Name
- keratin 42 pseudogene
There are no transcripts associated with this gene.
View Full Product Details