Product Details

SNP ID
rs942442
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:89132072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATTTGCTTTTTAATTTTAAACT[C/T]TTCTTAAATGAAACTGCAATAACAC
Phenotype
MIM: 600412 MIM: 612468
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
There are no transcripts associated with this gene.

Gene
GBP7
Gene Name
guanylate binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207398.2 2133 UTR 3 NP_997281.2

View Full Product Details