Product Details

SNP ID
rs1802046
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:77091898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAATGTATTTTTGAGAGGGAAAA[G/T]GTTGATAATACATTGTAAAATGAAA
Phenotype
MIM: 605087
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIGK PubMed Links

Gene Details

Gene
PIGK
Gene Name
phosphatidylinositol glycan anchor biosynthesis class K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005482.2 1709 UTR 3 NP_005473.1

View Full Product Details