Product Details

SNP ID
rs1043451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66284709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGCAGCTCCCAGAGTCAGGTA[A/G]AGCTGGAGACGCTGCCGGGGGACGG
Phenotype
MIM: 611288 MIM: 612565 MIM: 611484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182553.2 1057 Intron NP_872359.1
Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM151A
Gene Name
transmembrane protein 151A
There are no transcripts associated with this gene.

Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300861.1 1057 Silent Mutation CTC,CTT L218L NP_001287790.1
NM_020470.2 1057 Silent Mutation CTC,CTT L270L NP_065203.2
XM_005273720.3 1057 Missense Mutation TCT,TTT S300F XP_005273777.1
XM_017017139.1 1057 Missense Mutation TCT,TTT S312F XP_016872628.1
XM_017017140.1 1057 Silent Mutation CTC,CTT L282L XP_016872629.1
XM_017017141.1 1057 Intron XP_016872630.1

View Full Product Details