Product Details

SNP ID
rs10881623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89695258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCATTCTCATCTTCCCCAGA[C/T]TTGTTTAGATCTTCCTCAAATGTCT
Phenotype
MIM: 605498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ37201 PubMed Links

Gene Details

Gene
FLJ37201
Gene Name
tigger transposable element derived 2 pseudogene
There are no transcripts associated with this gene.

Gene
KIF20B
Gene Name
kinesin family member 20B
There are no transcripts associated with this gene.

View Full Product Details