Product Details

SNP ID
rs10951929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47786683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATGTGGAGGAAAAACCCTTC[C/T]GACATCTCTAAATTGCATCCACTGA
Phenotype
MIM: 609721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf69 PubMed Links

Gene Details

Gene
C7orf69
Gene Name
chromosome 7 open reading frame 69
There are no transcripts associated with this gene.

Gene
PKD1L1
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138295.3 Intron NP_612152.1
XM_017011798.1 Intron XP_016867287.1

View Full Product Details