Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_031910.3 | 3764 | UTR 3 | NP_114116.3 | ||
NM_182486.1 | 3764 | Intron | NP_872292.1 | ||
XM_011529857.1 | 3764 | UTR 3 | XP_011528159.1 | ||
XM_017028569.1 | 3764 | UTR 3 | XP_016884058.1 | ||
XM_017028570.1 | 3764 | UTR 3 | XP_016884059.1 | ||
XM_017028571.1 | 3764 | UTR 3 | XP_016884060.1 | ||
XM_017028572.1 | 3764 | UTR 3 | XP_016884061.1 | ||
XM_017028573.1 | 3764 | UTR 3 | XP_016884062.1 | ||
XM_017028574.1 | 3764 | UTR 3 | XP_016884063.1 |