Product Details

SNP ID
rs2267030
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:23790187 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTCAGAGCCCTGCAGGGCCTGCA[A/G]GGTAGTCATAGGGGGGAGTATTGTC
Phenotype
MIM: 185261 MIM: 601607
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MMP11 PubMed Links
Additional Information
For this assay, SNP(s) [rs34814300] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

Gene
SMARCB1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007468.2 Intron NP_001007469.1
NM_001317946.1 Intron NP_001304875.1
NM_003073.4 Intron NP_003064.2
XM_011530345.2 Intron XP_011528647.1

View Full Product Details