Product Details

SNP ID
rs1048635
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:117112236 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TTCTATCAGTCTCTTTAGAATGAAC[A/G]AAGGTCTGGGTCCTCTGGAAATCTC
Phenotype
MIM: 609318 MIM: 604718
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM45 PubMed Links

Gene Details

Gene
TRIM45
Gene Name
tripartite motif containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145635.1 2400 UTR 3 NP_001139107.1
NM_025188.3 2400 UTR 3 NP_079464.2
XM_011542199.2 2400 UTR 3 XP_011540501.1
Gene
TTF2
Gene Name
transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details