Product Details

SNP ID
rs1761468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:66304248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGTCAAGTATAGCAGTCCCAGC[A/C]CAAATCAATGACAAAAAAAAAAAAA
Phenotype
MIM: 603581
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCDH9 PubMed Links

Gene Details

Gene
PCDH9
Gene Name
protocadherin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318372.1 4813 UTR 3 NP_001305301.1
NM_001318373.1 4813 UTR 3 NP_001305302.1
NM_001318374.1 4813 Intron NP_001305303.1
NM_020403.4 4813 UTR 3 NP_065136.1
NM_203487.2 4813 UTR 3 NP_982354.1
XM_005266408.3 4813 Intron XP_005266465.1
XM_011535099.2 4813 Intron XP_011533401.1
XM_017020619.1 4813 UTR 3 XP_016876108.1
XM_017020620.1 4813 Intron XP_016876109.1
XM_017020621.1 4813 Intron XP_016876110.1

View Full Product Details