Product Details

SNP ID
rs951898
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:122097442 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATTTCTTTGGAAACAGAGGCAAAAA[A/G]GGAATTTTTAAAAAGGAGGTGGGGA
Phenotype
MIM: 615965 MIM: 610104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR100HG PubMed Links

Gene Details

Gene
MIR100HG
Gene Name
mir-100-let-7a-2 cluster host gene
There are no transcripts associated with this gene.

Gene
MIR125B1
Gene Name
microRNA 125b-1
There are no transcripts associated with this gene.

View Full Product Details