Product Details

SNP ID
rs894189
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44703425 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCCGAGCGAAGGGGACAACGTCAC[A/G]CTGGTCGTCCATGGGCTTTCGGGGG
Phenotype
MIM: 614591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM16 PubMed Links

Gene Details

Gene
CEACAM16
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039213.3 320 Silent Mutation ACA,ACG T38T NP_001034302.2
XM_017026795.1 320 Silent Mutation ACA,ACG T38T XP_016882284.1

View Full Product Details