Product Details

SNP ID
rs1046922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76026872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCATTGTTCTCAGGGAATGAC[A/T]TGAGAGCAAAGAAAAGGAACACTCA
Phenotype
MIM: 605335
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHLDA1 PubMed Links

Gene Details

Gene
PHLDA1
Gene Name
pleckstrin homology like domain family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007350.3 4488 UTR 3 NP_031376.3

View Full Product Details