Product Details

SNP ID
rs1673082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36750239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGAATTCTCCAATGTTGAATAAG[A/G]TGTGCAGACGGTCTAAAGGCCTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 864 Silent Mutation CAC,CAT H267H NP_001180481.1
NM_001267779.1 864 Silent Mutation CAC,CAT H235H NP_001254708.1

View Full Product Details