Product Details

SNP ID
rs1055394
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31681425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCTTAGAGTCACAAACTACCA[A/G]ACAGAAAAGGAACTGCAGGGGCATC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PRR14L PubMed Links

Gene Details

Gene
PRR14L
Gene Name
proline rich 14 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173566.2 12471 UTR 3 NP_775837.2
XM_006724219.3 12471 Intron XP_006724282.1
XM_006724220.3 12471 Intron XP_006724283.1
XM_006724221.2 12471 Intron XP_006724284.1
XM_011530080.2 12471 Intron XP_011528382.1
XM_011530081.2 12471 Intron XP_011528383.1
XM_017028736.1 12471 UTR 3 XP_016884225.1
XM_017028737.1 12471 Intron XP_016884226.1

View Full Product Details