Product Details

SNP ID
rs1507614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3512384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCAGGGGCACAAAGTCTCTATC[G/T]GTCCAGCTAGTCTGGCACACAGAGT
Phenotype
MIM: 608034 MIM: 607066
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPA PubMed Links

Gene Details

Gene
ASPA
Gene Name
aspartoacylase
There are no transcripts associated with this gene.

Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 4228 Intron NP_001164166.1
NM_001170696.1 4228 Intron NP_001164167.1
NM_001170697.1 4228 Intron NP_001164168.1
NM_001170698.1 4228 Intron NP_001164169.1
NM_001170699.1 4228 Intron NP_001164170.1
NM_001321336.1 4228 Intron NP_001308265.1
NM_001321337.1 4228 Intron NP_001308266.1
NM_032598.4 4228 Intron NP_115987.2
Gene
TRPV3
Gene Name
transient receptor potential cation channel subfamily V member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258205.1 4228 UTR 3 NP_001245134.1
NM_145068.3 4228 UTR 3 NP_659505.1

View Full Product Details