Product Details

SNP ID
rs954071
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:205792941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTGCTCTCCTGTCCTCTCTCT[C/T]TTTTTTTTTTTAAATTTTAGGCTCT
Phenotype
MIM: 610801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC41A1 PubMed Links

Gene Details

Gene
SLC41A1
Gene Name
solute carrier family 41 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173854.5 Intron NP_776253.3
XM_005245069.2 Intron XP_005245126.1

View Full Product Details