Product Details

SNP ID
rs1802944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88898987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGGCTGTTATCTCCAGCCTGG[C/G]GCTTCCGGGCCTTTCTTGAGGCCAC
Phenotype
MIM: 602281
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAPLN3 PubMed Links

Gene Details

Gene
HAPLN3
Gene Name
hyaluronan and proteoglycan link protein 3
There are no transcripts associated with this gene.

Gene
MFGE8
Gene Name
milk fat globule-EGF factor 8 protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114614.2 1722 UTR 3 NP_001108086.1
NM_001310319.1 1722 UTR 3 NP_001297248.1
NM_001310320.1 1722 UTR 3 NP_001297249.1
NM_001310321.1 1722 UTR 3 NP_001297250.1
NM_005928.3 1722 UTR 3 NP_005919.2
XM_017022206.1 1722 Intron XP_016877695.1

View Full Product Details