Product Details

SNP ID
rs1802121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24819543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCATGTCCTGCCCCACCAAGC[C/G]CTGTCTGAAGTGGAGCTTCCCCGCC
Phenotype
MIM: 600291 MIM: 611504
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCY3 PubMed Links

Gene Details

Gene
ADCY3
Gene Name
adenylate cyclase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320613.1 1602 UTR 3 NP_001307542.1
NM_004036.4 1602 UTR 3 NP_004027.2
XM_006711925.1 1602 UTR 3 XP_006711988.1
XM_011532489.1 1602 UTR 3 XP_011530791.1
XM_011532490.1 1602 UTR 3 XP_011530792.1
XM_011532491.1 1602 UTR 3 XP_011530793.1
XM_011532492.2 1602 UTR 3 XP_011530794.1
XM_011532493.1 1602 UTR 3 XP_011530795.1
XM_011532494.1 1602 UTR 3 XP_011530796.1
XM_011532496.1 1602 UTR 3 XP_011530798.1
XM_017003186.1 1602 UTR 3 XP_016858675.1
XM_017003187.1 1602 UTR 3 XP_016858676.1
XM_017003188.1 1602 UTR 3 XP_016858677.1
XM_017003189.1 1602 UTR 3 XP_016858678.1
XM_017003190.1 1602 UTR 3 XP_016858679.1
XM_017003191.1 1602 UTR 3 XP_016858680.1
XM_017003192.1 1602 UTR 3 XP_016858681.1
XM_017003193.1 1602 UTR 3 XP_016858682.1
Gene
CENPO
Gene Name
centromere protein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199803.2 1602 UTR 3 NP_001186732.1
NM_001322101.1 1602 UTR 3 NP_001309030.1
NM_024322.3 1602 UTR 3 NP_077298.1

View Full Product Details