Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320613.1 | 1815 | UTR 3 | NP_001307542.1 | ||
NM_004036.4 | 1815 | UTR 3 | NP_004027.2 | ||
XM_006711925.1 | 1815 | UTR 3 | XP_006711988.1 | ||
XM_011532489.1 | 1815 | UTR 3 | XP_011530791.1 | ||
XM_011532490.1 | 1815 | UTR 3 | XP_011530792.1 | ||
XM_011532491.1 | 1815 | UTR 3 | XP_011530793.1 | ||
XM_011532492.2 | 1815 | UTR 3 | XP_011530794.1 | ||
XM_011532493.1 | 1815 | UTR 3 | XP_011530795.1 | ||
XM_011532494.1 | 1815 | UTR 3 | XP_011530796.1 | ||
XM_011532496.1 | 1815 | UTR 3 | XP_011530798.1 | ||
XM_017003186.1 | 1815 | UTR 3 | XP_016858675.1 | ||
XM_017003187.1 | 1815 | UTR 3 | XP_016858676.1 | ||
XM_017003188.1 | 1815 | UTR 3 | XP_016858677.1 | ||
XM_017003189.1 | 1815 | UTR 3 | XP_016858678.1 | ||
XM_017003190.1 | 1815 | UTR 3 | XP_016858679.1 | ||
XM_017003191.1 | 1815 | UTR 3 | XP_016858680.1 | ||
XM_017003192.1 | 1815 | UTR 3 | XP_016858681.1 | ||
XM_017003193.1 | 1815 | UTR 3 | XP_016858682.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199803.2 | 1815 | UTR 3 | NP_001186732.1 | ||
NM_001322101.1 | 1815 | UTR 3 | NP_001309030.1 | ||
NM_024322.3 | 1815 | UTR 3 | NP_077298.1 |