Product Details

SNP ID
rs1157900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6640928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAAAAAGCCATGCGGATGGATT[A/G]TGGTCGTCAGAGGCTCCCGTAGTCT
Phenotype
MIM: 616967
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0753 PubMed Links

Gene Details

Gene
KIAA0753
Gene Name
KIAA0753
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014804.2 27 Intron NP_055619.2
XM_006721612.2 27 Intron XP_006721675.1
XM_011524090.2 27 Intron XP_011522392.1
XM_011524091.2 27 Intron XP_011522393.1
XM_011524095.2 27 Intron XP_011522397.1
XM_011524096.1 27 Intron XP_011522398.1
XM_011524098.2 27 Intron XP_011522400.1
XM_011524099.2 27 Intron XP_011522401.1
XM_011524100.2 27 Intron XP_011522402.1
XM_017025455.1 27 Intron XP_016880944.1
XM_017025456.1 27 Intron XP_016880945.1
XM_017025457.1 27 Intron XP_016880946.1
Gene
MED31
Gene Name
mediator complex subunit 31
There are no transcripts associated with this gene.

Gene
TXNDC17
Gene Name
thioredoxin domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032731.3 27 UTR 5 NP_116120.1

View Full Product Details