Product Details

SNP ID
rs1044809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3662993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGTGCACTTGCTGAGCTTCCA[A/G]ATGCGAATTAGTAAAAGCTAAATTC
Phenotype
MIM: 606272 MIM: 616484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031681.2 4321 UTR 3 NP_001026851.2
NM_004937.2 4321 UTR 3 NP_004928.2
XM_005256485.2 4321 Intron XP_005256542.1
XM_006721463.2 4321 Intron XP_006721526.1
XM_006721464.2 4321 Intron XP_006721527.1
XM_011523691.2 4321 Intron XP_011521993.1
XM_011523692.2 4321 Intron XP_011521994.1
XM_017024254.1 4321 Intron XP_016879743.1
XM_017024255.1 4321 Intron XP_016879744.1
XM_017024256.1 4321 Intron XP_016879745.1
XM_017024257.1 4321 Intron XP_016879746.1
XM_017024258.1 4321 Intron XP_016879747.1
Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204698.1 4321 UTR 3 NP_001191627.1
NM_014604.3 4321 UTR 3 NP_055419.1

View Full Product Details