Product Details

SNP ID
rs1044811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3662974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACGGTAACCAAAGTAAGGCTTG[G/T]GCACTTGCTGAGCTTCCAGATGCGA
Phenotype
MIM: 606272 MIM: 616484
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031681.2 4302 UTR 3 NP_001026851.2
NM_004937.2 4302 UTR 3 NP_004928.2
XM_005256485.2 4302 Intron XP_005256542.1
XM_006721463.2 4302 Intron XP_006721526.1
XM_006721464.2 4302 Intron XP_006721527.1
XM_011523691.2 4302 Intron XP_011521993.1
XM_011523692.2 4302 Intron XP_011521994.1
XM_017024254.1 4302 Intron XP_016879743.1
XM_017024255.1 4302 Intron XP_016879744.1
XM_017024256.1 4302 Intron XP_016879745.1
XM_017024257.1 4302 Intron XP_016879746.1
XM_017024258.1 4302 Intron XP_016879747.1
Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204698.1 4302 UTR 3 NP_001191627.1
NM_014604.3 4302 UTR 3 NP_055419.1

View Full Product Details