Product Details

SNP ID
rs1058636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67667981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCAGAGGACAGCCAGAGTGAG[C/G]CCTCACCATCTGCCAAACAGCACAA
Phenotype
MIM: 609377 MIM: 607484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
There are no transcripts associated with this gene.

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012984.2 601 Missense Mutation CCC,GCC P146A NP_001013002.2
XM_005255952.4 601 Missense Mutation CCC,GCC P146A XP_005256009.1
Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
GFOD2
Gene Name
glucose-fructose oxidoreductase domain containing 2
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
There are no transcripts associated with this gene.

View Full Product Details