Product Details
- SNP ID
-
rs1110657
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56794326 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTATATCTTGTCCTGCTTGGTCC[A/C]CAAAACTTACAGAGGAGAAATGAAA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf67
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75088376] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C17orf67
- Gene Name
- chromosome 17 open reading frame 67
View Full Product Details