Product Details

SNP ID
rs1047791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237574401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATATCTCAGGCGAAATGTCTCA[A/G]AATCTTCCTGCTCATTGGACAGAAA
Phenotype
MIM: 602663 MIM: 602206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRLH PubMed Links

Gene Details

Gene
PRLH
Gene Name
prolactin releasing hormone
There are no transcripts associated with this gene.

Gene
RAB17
Gene Name
RAB17, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022449.3 1803 UTR 3 NP_071894.1
XM_006712689.1 1803 UTR 3 XP_006712752.1
XM_017004693.1 1803 Intron XP_016860182.1
XM_017004694.1 1803 Intron XP_016860183.1

View Full Product Details