Product Details

SNP ID
rs2575651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108651634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAAGATTTACTTAGAACTGTGCT[A/G]CTGTGTACATTTAGATTTGGGGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSTC PubMed Links

Gene Details

Gene
OSTC
Gene Name
oligosaccharyltransferase complex non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267817.1 Intron NP_001254746.1
NM_001267818.1 Intron NP_001254747.1
NM_021227.3 Intron NP_067050.1

View Full Product Details