Product Details

SNP ID
rs3766
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30357270 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAGGGAACAAGGTGAAAGGTAT[C/G]GGTCCTGGTGAGACAAAAGCAGGGG
Phenotype
MIM: 604470 MIM: 613620
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CD2BP2 PubMed Links

Gene Details

Gene
CD2BP2
Gene Name
CD2 cytoplasmic tail binding protein 2
There are no transcripts associated with this gene.

Gene
TBC1D10B
Gene Name
TBC1 domain family member 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015527.3 2986 UTR 3 NP_056342.3
XM_011545789.1 2986 UTR 3 XP_011544091.1
XM_011545790.2 2986 UTR 3 XP_011544092.1

View Full Product Details