Product Details

SNP ID
rs1466685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49722606 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCTCCTACACACCTGGATGCGC[C/T]GCAGCACTGCAGGGCTCAGGATGTA
Phenotype
MIM: 615691 MIM: 615320 MIM: 606991 MIM: 614472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMIGO3 PubMed Links

Gene Details

Gene
AMIGO3
Gene Name
adhesion molecule with Ig like domain 3
There are no transcripts associated with this gene.

Gene
GMPPB
Gene Name
GDP-mannose pyrophosphorylase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013334.3 799 Missense Mutation CAG,CGG Q184R NP_037466.2
NM_021971.2 799 Missense Mutation CAG,CGG Q184R NP_068806.1
Gene
IP6K1
Gene Name
inositol hexakisphosphate kinase 1
There are no transcripts associated with this gene.

Gene
RNF123
Gene Name
ring finger protein 123
There are no transcripts associated with this gene.

View Full Product Details