Product Details

SNP ID
rs1057363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49684015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTCCTCCCCAAGGCATATGGC[A/G]TCAAGGCTGGGCCTAACCCAGTCTC
Phenotype
MIM: 102645 MIM: 142408 MIM: 614472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APEH PubMed Links

Gene Details

Gene
APEH
Gene Name
acylaminoacyl-peptide hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001640.3 3057 Intron NP_001631.3
XM_005265097.2 3057 Intron XP_005265154.1
XM_005265098.4 3057 Intron XP_005265155.1
XM_011533658.2 3057 Intron XP_011531960.1
XM_011533659.1 3057 Intron XP_011531961.1
XM_011533660.2 3057 Intron XP_011531962.1
XM_011533661.2 3057 Intron XP_011531963.1
XM_011533662.2 3057 Intron XP_011531964.1
XM_011533663.2 3057 Intron XP_011531965.1
XM_017006285.1 3057 Intron XP_016861774.1
XM_017006286.1 3057 Intron XP_016861775.1
Gene
MST1
Gene Name
macrophage stimulating 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020998.3 3057 UTR 3 NP_066278.3
XM_006713166.1 3057 Intron XP_006713229.1
XM_011533732.1 3057 Intron XP_011532034.1
XM_011533737.2 3057 Intron XP_011532039.1
XM_011533738.2 3057 Intron XP_011532040.1
XM_017006460.1 3057 Intron XP_016861949.1
XM_017006461.1 3057 Intron XP_016861950.1
XM_017006462.1 3057 UTR 3 XP_016861951.1
XM_017006463.1 3057 Intron XP_016861952.1
XM_017006464.1 3057 UTR 3 XP_016861953.1
XM_017006465.1 3057 Intron XP_016861954.1
XM_017006466.1 3057 Intron XP_016861955.1
XM_017006467.1 3057 Intron XP_016861956.1
XM_017006468.1 3057 Intron XP_016861957.1
Gene
RNF123
Gene Name
ring finger protein 123
There are no transcripts associated with this gene.

View Full Product Details