Product Details

SNP ID
rs3864652
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117076544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAGTATGCGAGCAAACAAGGTA[A/G]GAACTGGAGTGCATGAGCGCTGGAG
Phenotype
MIM: 600833
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ST7 PubMed Links

Gene Details

Gene
ST7
Gene Name
suppression of tumorigenicity 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018412.3 Intron NP_060882.2
NM_021908.2 Intron NP_068708.1
Gene
ST7-AS2
Gene Name
ST7 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details